2-Formyl-4-(4-methylphenyl)phenol ≥97%

Supplier: AOBChem USA

Warning

35030-500MG 35030-1G 35030-5G 35030-25G
77325-768EA 145.23 USD
77325-768 77345-126 77362-116 77385-124
2-Formyl-4-(4-methylphenyl)phenol ≥97%
2-Formyl-4-(4-methylphenyl)phenol

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₂
MW: 212.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06802128
CAS Number: 893737-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR