3-Fluorobenzene-1,2-dicarbaldehyde ≥97%

Supplier: AOBChem USA

Warning

34984-500MG 34984-1G 34984-5G 34984-25G
77325-766EA 279.13 USD
77325-766 77349-432 77362-114 77385-122
3-Fluorobenzene-1,2-dicarbaldehyde ≥97%
3-Fluorobenzene-1,2-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅FO₂
MW: 152.123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12031830
CAS Number: 161747-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR