5,6-Dimethoxynicotinic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
34968-500MG
34968-1G
34968-5G
34968-25G
77325-764EA
222.48
USD
77325-764
77349-430
77362-112
77385-120
5,6-Dimethoxynicotinic acid ≥95%
5,6-Dimethoxynicotinic acid
Precaución: for Research Use Only
Formula:
C₈H₉NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD12401642 CAS Number: 76470-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...