2-Bromo-6-chloro-3-(trifluoromethyl)benzyl alcohol ≥95%
Supplier: AOBChem USA
|
Warning
|
34956-500MG
34956-1G
34956-5G
34956-25G
77325-760EA
182.31
USD
77325-760
77346-926
77362-108
77385-116
2-Bromo-6-chloro-3-(trifluoromethyl)benzyl alcohol ≥95%
2-Bromo-6-chloro-3-(trifluoromethyl)benzyl alcohol
Precaución: for Research Use Only
Formula:
C₈H₅BrClF₃O MW: 289.477 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34755975 CAS Number: 2091848-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...