3-Benzyloxy-4-methylbenzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

34925-500MG 34925-1G 34925-25G 34925-5G
77325-756EA 297.67 USD
77325-756 77349-424 77385-112 77373-676
3-Benzyloxy-4-methylbenzyl alcohol ≥95%
3-Benzyloxy-4-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₂
MW: 228.286 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658149
CAS Number: 916792-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR