2-Bromo-3-(benzyloxy)-6-fluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

34921-500MG 34921-1G 34921-5G 34921-25G
77325-754EA 97.85 USD
77325-754 77342-656 77362-104 77373-674
2-Bromo-3-(benzyloxy)-6-fluorobenzoic acid ≥95%
2-Bromo-3-(benzyloxy)-6-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrFO₃
MW: 325.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707003
CAS Number: 2432848-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR