1-(Benzyloxy)-6-bromo-2-chloro-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

34920-500MG 34920-1G 34920-5G 34920-25G
77325-752EA 201.88 USD
77325-752 77349-422 77362-102 77385-110
1-(Benzyloxy)-6-bromo-2-chloro-3-methylbenzene ≥95%
1-(Benzyloxy)-6-bromo-2-chloro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrClO
MW: 311.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31805780
CAS Number: 2244107-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR