1-(4-Phenylthiophen-2-yl)ethan-1-one ≥97%

Supplier: AOBChem USA

Warning

34855-500MG 34855-1G 34855-5G 34855-25G
77325-748EA 95.79 USD
77325-748 77342-542 77362-098 77373-668
1-(4-Phenylthiophen-2-yl)ethan-1-one ≥97%
1-(4-Phenylthiophen-2-yl)ethan-1-one

Precaución: for Research Use Only

Formula: C₁₂H₁₀OS
Storage Temperature: Ambient
MDL Number: MFCD01927465
CAS Number: 26170-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR