2-Fluoro-5-isopropoxypyridin-3-ol ≥95%

Supplier: AOBChem USA

Warning

34784-500MG 34784-1G 34784-5G
77339-052EA 547.96 USD
77339-052 77349-412 77373-660
2-Fluoro-5-isopropoxypyridin-3-ol ≥95%
2-Fluoro-5-isopropoxypyridin-3-ol

Precaución: for Research Use Only

Formula: C₈H₁₀FNO₂
Storage Temperature: Ambient
MDL Number: MFCD31696109
CAS Number: 2230481-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR