3-Chloro-6-ethoxyphenyl methyl sulfide ≥95%

Supplier: AOBChem USA

Warning

34779-500MG 34779-1G 34779-5G 34779-25G
77325-734EA 243.08 USD
77325-734 77349-408 77362-082 77385-096
3-Chloro-6-ethoxyphenyl methyl sulfide ≥95%
3-Chloro-6-ethoxyphenyl methyl sulfide

Precaución: for Research Use Only

Formula: C₉H₁₁ClOS
Storage Temperature: Ambient
MDL Number: MFCD22370313
CAS Number: 1443329-44-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR