2,4-Bis(trifluoromethyl)thioanisole ≥97%

Supplier: AOBChem USA

Warning

34777-500MG 34777-1G 34777-5G 34777-25G
77325-732EA 100.94 USD
77325-732 77342-914 77362-080 77373-658
2,4-Bis(trifluoromethyl)thioanisole ≥97%
2,4-Bis(trifluoromethyl)thioanisole

Precaución: for Research Use Only

Formula: C₉H₆F₆S
MW: 260.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28733246
CAS Number: 393-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR