2-Bromo-1-butoxy-4-chlorobenzene ≥97%

Supplier: AOBChem USA

Warning

34765-500MG 34765-1G 34765-5G 34765-25G
77325-728EA 174.07 USD
77325-728 77346-440 77362-076 77385-092
2-Bromo-1-butoxy-4-chlorobenzene ≥97%
2-Bromo-1-butoxy-4-chlorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrClO
MW: 263.559 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16335684
CAS Number: 1225548-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR