[2,4-Difluoro-3-(propan-2-yloxy)phenyl]methanol ≥97%

Supplier: AOBChem USA

Warning

34763-500MG 34763-1G 34763-5G 34763-25G
77325-726EA 233.81 USD
77325-726 77349-404 77362-074 77385-090
[2,4-Difluoro-3-(propan-2-yloxy)phenyl]methanol ≥97%
[2,4-Difluoro-3-(propan-2-yloxy)phenyl]methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₂O₂
MW: 202.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31696107
CAS Number: 2230481-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR