4-Butoxy-3,5-dichlorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

34755-500MG 34755-1G 34755-5G 34755-25G
77325-718EA 260.59 USD
77325-718 77349-400 77362-068 77385-086
4-Butoxy-3,5-dichlorobenzoic acid ≥97%
4-Butoxy-3,5-dichlorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD08690662
CAS Number: 41490-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR