(5-Fluoro-2-isopropoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

34748-500MG 34748-1G 34748-5G
77325-716EA 336.81 USD
77325-716 77349-398 77373-650
(5-Fluoro-2-isopropoxyphenyl)methanol ≥95%
(5-Fluoro-2-isopropoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO₂
MW: 184.207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11639982
CAS Number: 610797-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR