1-Benzyloxy-6-bromo-2,3-difluorobenzene ≥95%

Supplier: AOBChem USA

Warning

34747-500MG 34747-1G 34747-5G 34747-25G
77325-714EA 233.81 USD
77325-714 77349-396 77362-066 77385-084
1-Benzyloxy-6-bromo-2,3-difluorobenzene ≥95%
1-Benzyloxy-6-bromo-2,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉BrF₂O
MW: 299.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31696503
CAS Number: 2230481-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR