4-Ethoxy-2-(trifluoromethyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

34722-500MG 34722-1G 34722-5G 34722-25G
77325-710EA 187.46 USD
77325-710 77349-394 77362-062 77385-082
4-Ethoxy-2-(trifluoromethyl)benzaldehyde ≥95%
4-Ethoxy-2-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂
MW: 218.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24141328
CAS Number: 1502383-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR