4-Ethoxy-2-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

34721-500MG 34721-1G 34721-5G 34721-25G
77325-708EA 200.85 USD
77325-708 77349-392 77362-060 77385-080
4-Ethoxy-2-(trifluoromethyl)benzoic acid ≥97%
4-Ethoxy-2-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD16852853
CAS Number: 1340229-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR