(3-Chloro-4-phenoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

34712-500MG 34712-1G 34712-5G 34712-25G
77325-704EA 182.31 USD
77325-704 77346-924 77362-056 77385-076
(3-Chloro-4-phenoxyphenyl)methanol ≥95%
(3-Chloro-4-phenoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClO₂
MW: 234.678 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24056421
CAS Number: 1228582-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR