3-(4-Fluoro-3-methylphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

34694-500MG 34694-1G 34694-5G
77325-700EA 353.29 USD
77325-700 77349-382 77373-642
3-(4-Fluoro-3-methylphenyl)pyridine ≥97%
3-(4-Fluoro-3-methylphenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀FN
MW: 187.213 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15477315
CAS Number: 1214328-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR