3-(4-Chlorophenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

34690-500MG 34690-1G 34690-5G 34690-25G
77325-698EA 202.91 USD
77325-698 77349-380 77362-052 77385-072
3-(4-Chlorophenyl)pyridine ≥97%
3-(4-Chlorophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₈ClN
MW: 189.641 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07644611
CAS Number: 5957-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR