(3,5-Dibromo-4-butoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

34658-500MG 34658-1G 34658-5G 34658-25G
77325-692EA 168.92 USD
77325-692 77346-250 77362-048 77385-068
(3,5-Dibromo-4-butoxyphenyl)methanol ≥97%
(3,5-Dibromo-4-butoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₄Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD31619522
CAS Number: 1697692-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR