3,5-Dibromo-4-ethoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

34656-500MG 34656-1G 34656-5G 34656-25G
77325-690EA 91.67 USD
77325-690 77342-426 77362-046 77373-628
3,5-Dibromo-4-ethoxybenzyl alcohol ≥97%
3,5-Dibromo-4-ethoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₀Br₂O₂
MW: 309.983 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03862354
CAS Number: 664319-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR