2-Bromo-6-chloro-3-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

34618-500MG 34618-1G 34618-5G 34618-25G
77325-682EA 134.93 USD
77325-682 77344-448 77362-038 77385-064
2-Bromo-6-chloro-3-methylbenzyl alcohol ≥97%
2-Bromo-6-chloro-3-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD31619514
CAS Number: 2091935-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR