4-Acetyl-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

34584-500MG 34584-1G 34584-5G
77336-236EA 384.19 USD
77336-236 77349-360 77373-610
4-Acetyl-2-methoxybenzaldehyde ≥97%
4-Acetyl-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD24681392
CAS Number: 109142-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR