2-(4-Methoxyphenyl)pyridine-5-boronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

34558-500MG 34558-1G 34558-5G 34558-25G
77325-676EA 169.95 USD
77325-676 77346-340 77362-030 77385-060
2-(4-Methoxyphenyl)pyridine-5-boronic acid pinacol ester ≥95%
2-(4-Methoxyphenyl)pyridine-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₈H₂₂BNO₃
Storage Temperature: Ambient
MDL Number: MFCD22566473
CAS Number: 1374263-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR