5-Bromo-2-(4-methoxyphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

34556-500MG 34556-1G 34556-5G 34556-25G
77325-674EA 263.68 USD
77325-674 77349-358 77362-028 77385-058
5-Bromo-2-(4-methoxyphenyl)pyridine ≥97%
5-Bromo-2-(4-methoxyphenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO
MW: 264.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14707260
CAS Number: 88345-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR