3-Fluoro-5-(methylthio)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

34549-500MG 34549-1G 34549-5G
77338-122EA 466.59 USD
77338-122 77349-356 77373-608
3-Fluoro-5-(methylthio)phenylboronic acid pinacol ester ≥97%
3-Fluoro-5-(methylthio)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₂S
Storage Temperature: Ambient
MDL Number: MFCD31619500
CAS Number: 2762075-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR