2-Iodo-4-methylthiophene-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

34543-500MG 34543-1G 34543-5G 34543-25G
77325-670EA 161.71 USD
77325-670 77345-836 77362-024 77385-054
2-Iodo-4-methylthiophene-3-carboxylic acid ≥97%
2-Iodo-4-methylthiophene-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅IO₂S
MW: 268.072 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564024
CAS Number: 2734774-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR