2,3-Dichlorobiphenyl (PCB No. 5) ≥97%

Supplier: AOBChem USA

Warning

34500-500MG 34500-1G 34500-5G
77336-456EA 388.31 USD
77336-456 77349-352 77373-604
2,3-Dichlorobiphenyl (PCB No. 5) ≥97%
2,3-Dichlorobiphenyl (PCB No. 5)

Precaución: for Research Use Only

Formula: C₁₂H₈Cl₂
Storage Temperature: Ambient
MDL Number: MFCD00055244
CAS Number: 16605-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR