2-Fluoro-4-methyl-5-phenylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

34483-500MG 34483-1G 34483-5G
77338-324EA 477.92 USD
77338-324 77349-350 77373-602
2-Fluoro-4-methyl-5-phenylbenzaldehyde ≥95%
2-Fluoro-4-methyl-5-phenylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD31619494
CAS Number: 2221812-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR