3-(4-Methoxyphenyl)thiophene ≥95%

Supplier: AOBChem USA

Warning

34477-500MG 34477-1G 34477-5G 34477-25G
77325-664EA 201.88 USD
77325-664 77349-346 77362-018 77385-048
3-(4-Methoxyphenyl)thiophene ≥95%
3-(4-Methoxyphenyl)thiophene

Precaución: for Research Use Only

Formula: C₁₁H₁₀OS
Storage Temperature: Ambient
MDL Number: MFCD02260246
CAS Number: 82437-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR