2-Chloro-4-fluoro-6-iodoaniline ≥97%

Supplier: AOBChem USA

Warning

34469-500MG 34469-1G 34469-5G 34469-25G
77325-662EA 202.91 USD
77325-662 77349-344 77362-016 77385-046
2-Chloro-4-fluoro-6-iodoaniline ≥97%
2-Chloro-4-fluoro-6-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₄ClFIN
MW: 271.459 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23132248
CAS Number: 1005196-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR