5-Chloro-4-fluoro-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

34375-500MG 34375-1G 34375-25G 34375-5G
77325-656EA 299.73 USD
77325-656 77349-338 77385-040 77373-594
5-Chloro-4-fluoro-2-methoxybenzaldehyde ≥97%
5-Chloro-4-fluoro-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20485230
CAS Number: 889858-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR