2,3-Dichloro-6-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

34374-500MG 34374-1G 34374-5G 34374-25G
77325-654EA 276.04 USD
77325-654 77349-336 77362-008 77385-038
2,3-Dichloro-6-methoxybenzyl alcohol ≥97%
2,3-Dichloro-6-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₈Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD28099829
CAS Number: 1823363-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR