1,2-Dibromo-3-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

34360-500MG 34360-1G 34360-5G 34360-25G
77325-652EA 198.79 USD
77325-652 77349-334 77362-006 77385-036
1,2-Dibromo-3-iodobenzene ≥95%
1,2-Dibromo-3-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃Br₂I
Storage Temperature: Ambient
MDL Number: MFCD13194295
CAS Number: 1191934-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR