2-(3,4-Dichloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

34329-500MG 34329-1G 34329-5G
77338-922EA 531.48 USD
77338-922 77349-326 77373-586
2-(3,4-Dichloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(3,4-Dichloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₂H₁₄BCl₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD22494820
CAS Number: 1165935-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR