3,6-Difluoro-2-methylphenol ≥97%

Supplier: AOBChem USA

Warning

34282-500MG 34282-1G 34282-5G
77336-776EA 394.49 USD
77336-776 77349-318 77373-576
3,6-Difluoro-2-methylphenol ≥97%
3,6-Difluoro-2-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆F₂O
MW: 144.119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18394890
CAS Number: 1046781-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR