Methyl 3-isopropyl-2-methoxybenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
34276-500MG
34276-1G
34276-5G
34276-25G
77325-646EA
122.57
USD
77325-646
77343-986
77362-002
77373-574
Methyl 3-isopropyl-2-methoxybenzoate ≥97%
Methyl 3-isopropyl-2-methoxybenzoate
Precaución: for Research Use Only
Formula:
C₁₂H₁₆O₃ MW: 208.254 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27930917 CAS Number: 87188-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...