3,4-Difluoro-2-(methylsulfanyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

34255-500MG 34255-1G 34255-5G 34255-25G
77325-644EA 120.51 USD
77325-644 77343-680 77362-000 77373-570
3,4-Difluoro-2-(methylsulfanyl)benzoic acid ≥97%
3,4-Difluoro-2-(methylsulfanyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂S
MW: 204.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24483524
CAS Number: 153556-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR