2-Bromo-3-fluoro-5-methylphenol ≥97%

Supplier: AOBChem USA

Warning

34115-500MG 34115-1G 34115-5G
77325-632EA 318.27 USD
77325-632 77349-310 77373-562
2-Bromo-3-fluoro-5-methylphenol ≥97%
2-Bromo-3-fluoro-5-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆BrFO
MW: 205.024 g/mol
Storage Temperature: Ambient
CAS Number: 1782495-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR