Ethyl 3,5-dichloro-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

34100-500MG 34100-1G 34100-5G
77325-630EA 313.12 USD
77325-630 77349-308 77373-560
Ethyl 3,5-dichloro-2-methylbenzoate ≥97%
Ethyl 3,5-dichloro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂O₂
MW: 233.091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24323017
CAS Number: 1552579-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR