1,5-Dichloro-3-iodo-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

34095-500MG 34095-1G 34095-5G 34095-25G
77325-628EA 160.68 USD
77325-628 77345-778 77361-984 77385-024
1,5-Dichloro-3-iodo-2-methylbenzene ≥95%
1,5-Dichloro-3-iodo-2-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂I
MW: 286.925 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11865414
CAS Number: 1215795-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR