4-tert-Butylcarbamoyl-2-chlorobenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

34089-500MG 34089-1G 34089-25G 34089-5G
77325-624EA 297.67 USD
77325-624 77349-302 77385-020 77373-554
4-tert-Butylcarbamoyl-2-chlorobenzeneboronic acid ≥97%
4-tert-Butylcarbamoyl-2-chlorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BClNO₃
MW: 255.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22125091
CAS Number: 2096336-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR