2,3-Dichloro-1,4-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

34074-500MG 34074-1G 34074-25G 34074-5G
77325-620EA 296.64 USD
77325-620 77349-300 77385-016 77373-552
2,3-Dichloro-1,4-dimethylbenzene ≥97%
2,3-Dichloro-1,4-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈Cl₂
MW: 175.055 g/mol
Storage Temperature: Ambient
CAS Number: 34840-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR