Ethyl 2-chloro-3-fluoro-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

34056-500MG 34056-1G 34056-5G
77339-050EA 547.96 USD
77339-050 77349-298 77373-550
Ethyl 2-chloro-3-fluoro-5-methylbenzoate ≥97%
Ethyl 2-chloro-3-fluoro-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO₂
MW: 216.637 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751854
CAS Number: 1805226-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR