Methyl 2-chloro-3-fluoro-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

34055-500MG 34055-1G 34055-5G
77339-048EA 547.96 USD
77339-048 77349-296 77373-548
Methyl 2-chloro-3-fluoro-5-methylbenzoate ≥97%
Methyl 2-chloro-3-fluoro-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD28751875
CAS Number: 1805454-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR