2-Chloro-3-fluoro-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

34054-500MG 34054-1G 34054-5G
77325-618EA 346.08 USD
77325-618 77349-294 77373-546
2-Chloro-3-fluoro-5-methylbenzoic acid ≥97%
2-Chloro-3-fluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23712096
CAS Number: 1427373-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR