2-Chloro-3-fluoro-5-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

34053-500MG 34053-1G 34053-5G
77339-046EA 547.96 USD
77339-046 77349-292 77373-544
2-Chloro-3-fluoro-5-methylbenzyl alcohol ≥97%
2-Chloro-3-fluoro-5-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₈ClFO
Storage Temperature: Ambient
MDL Number: MFCD28751963
CAS Number: 1807276-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR