2-(3,4-Difluorophenyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

34039-500MG 34039-1G 34039-5G 34039-25G
77325-614EA 218.36 USD
77325-614 77349-286 77361-980 77385-014
2-(3,4-Difluorophenyl)pyridine ≥95%
2-(3,4-Difluorophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₇F₂N
MW: 191.177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07775641
CAS Number: 387831-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR